missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROS Antibody (ROS1/8268R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20759JF635
This item is not returnable.
View return policy
Description
ROS Monoclonal antibody specifically detects ROS in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ROS | |
| Monoclonal | |
| Janelia Fluor 635 | |
| proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1, receptor tyrosine kinase, c-ros-1, MCF3, ROS | |
| Recombinant fragment corresponding to cytoplasmic domain of human ROS protein | |
| 0.1 mL | |
| Protein Kinase | |
| 6098 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| ROS1/8268R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction