missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RORB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17635-1-AP-150UL
This item is not returnable.
View return policy
Description
Retinoid-related orphan receptor beta (ROR beta), a NR1 thyroid hormone receptor, affects the development and functioning of the central nervous system, including sensory input integration and circadian timing. Disruption of ROR beta in mice results in juvenile ataxia, retinal degeneration, circadian activity abnormalities, and delayed onset of male fertility. ROR beta binds to the monomeric response elements containing the sequence AnnTAGGTCA as a monomer. An alternatively spliced isoform, ROR beta2, has been isolated from rats, and the expression of ROR beta2 is confined to the pineal gland and retina. ROR beta2 shares common DNA- and putative ligand-binding domains with wild-type ROR beta but is characterized by a different amino-terminal domain. ROR beta expression has been documented in various regions of the mouse brain.Specifications
| RORB | |
| Polyclonal | |
| Unconjugated | |
| RORB | |
| NR1F2, Nuclear receptor ROR beta, Nuclear receptor RZR beta, RAR related orphan receptor B, ROR BETA, RORB, RZR BETA, RZRB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 225998, 309288, 6096 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P45446, Q8R1B8, Q92753 | |
| RORB | |
| RORB Fusion Protein Ag11910 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction