missing translation for 'onlineSavingsMsg'
Learn More

ROPN1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12565-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890334

  • 1281.92 NOK / 20µL
Estimated Shipment: 20-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Principal Names: ROPN1L; Ropporin-1 like; ASP; AKAP-associated sperm protein; Official Gene Symbol: ROPN1L Gene ID: 83853 (human) Gene Map Locus: 5p15.2 (human) AKAPs act as scaffolding proteinscoordinating phosphatase/kinase signaling at the cytoskeleton. They interact with PKAs and anchor them to specific sub-cellular compartments and participate in multiple cellular processes including growth, metabolism and differentiation. ASP, a 26 kDa protein, is a novel AKAP-interacting protein. It interacts with the amphipathic helix of AKAP3 and facilitates AKAP3-PKA interactions. It consists of a conserved N-terminal AKAP-binding domain and a central cysteine-rich Fz domain. It is specifically expressed in the sperms and might play a significant role in sperm motility and also facilitate cytoskeletal reorganization during spermiogenesis.
Specifications

Specifications

ROPN1L
Polyclonal
Unconjugated
ROPN1L
AKAP associated sperm protein, ASP, ROPN1 like protein, ROPN1L, ropporin 1 like, Ropporin 1 like protein
Rabbit
Antigen Affinity Chromatography
RUO
252967, 685646, 83853
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96C74, Q9EQ00
Ropn1l
ROPN1L Fusion Protein Ag3178
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers