missing translation for 'onlineSavingsMsg'
Learn More

Ropivacaine hydrochloride, MedChemExpress

Product Code. p-200096384
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16454397

Brand: Medchem Express HYB0563B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ropivacaine hydrochloride is a potent sodium channel blocker and blocks impulse conduction via reversible inhibition of sodium ion influx in nerve fibrese. Ropivacaine is also an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. Ropivacaine is widely used for neuropathic pain management in vivo.

Ropivacaine hydrochloride is a potent sodium channel blocker and blocks impulse conduction via reversible inhibition of sodium ion influx in nerve fibrese. Ropivacaine is also an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. Ropivacaine is widely used for neuropathic pain management in vivo.

Specifications

Chemical Name or Material Ropivacaine hydrochloride
CAS 98717-15-8
Color White
Purity Grade Notes Research
Molecular Formula C17H27ClN2O
Quantity 50 mg
Solubility Information DMSO : 10 mg/mL (32.17 mM; Need ultrasonic) ∣H2O : 10 mg/mL (32.17 mM; Need ultrasonic)
SMILES [H]Cl.O=C([C@H]1N(CCC)CCCC1)NC2=C(C)C=CC=C2C
Molecular Weight (g/mol) 310.86
Formula Weight 310.86
Percent Purity 98.66%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.