Learn More
Brady™ Roll Markers
Brand: Brady™ 226383
Additional Details : Weight : 0.20000kg
Specifications
European Directives 67/548 and 88/379 | |
White on Blue | |
Polyester | |
Air | |
French | |
505 Pk. | |
-40°C to +140°C | |
0.119 mm | |
33 m x 100 mm x 60 mm | |
33 m | |
100 mm | |
0.31 in. | |
8 mm |
Permanent Acrylic | |
Roll | |
Pipe Marker | |
Pipe and valve marking | |
505 Markers/Roll | |
-40°F to +284°F | |
0.0047 in. | |
33 m x 100 mm | |
108.27 ft. | |
3.94 in. | |
2.36 in. | |
60 mm |