Learn More
Brady™ Roll Markers
Product Code. 11795573
Change view
Click to view available options
Width (Metric):
100 mm
Unit Size:
1 roll
Product Code. Width (Metric) unitSize
11795573 100 mm 1 roll
1 options
Product Code. 11795573
Brand: Brady™ 226903
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance European Directives 67/548 and 88/379
Color Orange
Material Polyester
Product Type Roll Marker
Display Text Gas
For Use With (Application) Used for Pipemarkers and safety signs
Quantity 505/Pk.
Length (Metric) 33 m
Width (Metric) 100 mm
Height (Metric) 100 mm
Height (Metric) Character 8 mm