Learn More
Rolipram, MedChemExpress
Product Code. 16473217
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16473217
Brand: Medchem Express HY16900
MedChemExpress Rolipram is a selective phosphodiesterases PDE4 inhibitor with IC50s of 3 nM, 130 nM and 240 nM for PDE4A, PDE4B, and PDE4D, respectively.
Rolipram is a selective phosphodiesterases PDE4 inhibitor with IC50s of 3 nM, 130 nM and 240 nM for PDE4A, PDE4B, and PDE4D, respectively.
Specifications
Chemical Name or Material Rolipram
CAS 61413-54-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H21NO3
Quantity 50 mg
Synonym (R,S)-Rolipram SB 95952 ZK 62711
Solubility Information DMSO : ≥ 41 mg/mL (148.91 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1NCC(C2=CC=C(OC)C(OC3CCCC3)=C2)C1
Molecular Weight (g/mol) 275.34
Formula Weight 275.34
Percent Purity 98.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.