Learn More
Rogo Sampaic™ Glass Coverslips
Product Code. 11707394
Change view
Click to view available options
Length (Metric):
20 mm
Width (Metric):
20 mm
Quantity:
200/Pk.
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Quantity Unit Size
11707394 20 mm 20 mm 200/Pk. Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11707394 Supplier Rogo Sampaic™ Supplier No. BPD017
  • ONLINE EXCLUSIVE PRICE
    49.90 NOK
    45.59 NOK / Pack of 200
    Save 4.31 NOK
    8% Off
Estimated Shipment: 26-11-2026
Preserve and protect specimens on slides. Rogo Sampaic™ Glass Coverslips are manufactured from borosilicate glass for excellent transparency and prevention of color distortion.
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Product Type Cover Slide
Shape Square
Material Borosilicate Glass
Length (Metric) 20 mm
Width (Metric) 20 mm
Dimensions (L x W) 20 x 20 mm
For Use With (Application) Slide microscopy
Certifications ISO 8255
Quantity 200/Pk.