missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
4260.00 NOK
Specifications
| Antigen | ROD1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ROD1 Polyclonal specifically detects ROD1 in Human samples. It is validated for Western Blot.Specifications
| ROD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp781I1117, fission yeast differentiation regulator, PTBP3, regulator of differentiation (in S. pombe) 1, regulator of differentiation 1, Rod1, ROD1 regulator of differentiation 1 (S. pombe) | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human ROD1 (NP_001157260.1). Peptide sequence RATLSKHQAVQLPREGQEDQGLTKDFSNSPLHRFKKPGSKNFQNIFPPSA | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 9991 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title