missing translation for 'onlineSavingsMsg'
Learn More

Roblitinib, MedChemExpress

Product Code. p-200095891
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16493418 10 mg 10mg
16464646 100 mg 100mg
16456147 5 mg 5mg
16449946 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16493418

Brand: Medchem Express HY101568

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Roblitinib (FGF-401) is an orally active and highly selective FGFR4 inhibitor with an IC50 of 1.9 nM. Roblitinib has antitumor activity.

Roblitinib (FGF-401) is an orally active and highly selective FGFR4 inhibitor with an IC50 of 1.9 nM. Roblitinib has antitumor activity.

Specifications

Chemical Name or Material Roblitinib
CAS 1708971-55-4
Color Light Nude
Purity Grade Notes Research
Molecular Formula C25H30N8O4
Quantity 10 mg
Synonym FGF-401
Solubility Information DMSO : 5.2 mg/mL (10.27 mM; Need ultrasonic)
SMILES CN1CC(N(CC2=CC(CCCN3C(NC4=CC(NCCOC)=C(C#N)C=N4)=O)=C3N=C2C=O)CC1)=O
Molecular Weight (g/mol) 506.56
Formula Weight 506.56
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.