Learn More
Roblitinib, MedChemExpress
Product Code. 16493418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493418 10 mg 10mg
16464646 100 mg 100mg
16456147 5 mg 5mg
16449946 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16493418 Supplier Medchem Express Supplier No. HY101568
MedChemExpress Roblitinib (FGF-401) is an orally active and highly selective FGFR4 inhibitor with an IC50 of 1.9 nM. Roblitinib has antitumor activity.
Roblitinib (FGF-401) is an orally active and highly selective FGFR4 inhibitor with an IC50 of 1.9 nM. Roblitinib has antitumor activity.
Specifications
Chemical Name or Material Roblitinib
CAS 1708971-55-4
Color Light Nude
Purity Grade Notes Research
Molecular Formula C25H30N8O4
Quantity 10 mg
Synonym FGF-401
Solubility Information DMSO : 5.2 mg/mL (10.27 mM; Need ultrasonic)
SMILES CN1CC(N(CC2=CC(CCCN3C(NC4=CC(NCCOC)=C(C#N)C=N4)=O)=C3N=C2C=O)CC1)=O
Molecular Weight (g/mol) 506.56
Formula Weight 506.56
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease