Learn More
RO9021, MedChemExpress
Product Code. 16497198
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16497198 10 mg 10mg
16481649 1 mg 1mg
16479727 5 mg 5mg
16437186 100 mg 100mg
16403227 50 mg 50mg
5 options
Product Code. 16497198
Brand: Medchem Express HY16902
MedChemExpress RO9021 is an orally bioavailable, novel ATP-competitive inhibitor of SYK, with an average IC50 of 5.6 nM.
RO9021 is an orally bioavailable, novel ATP-competitive inhibitor of SYK, with an average IC50 of 5.6 nM.
Specifications
Chemical Name or Material RO9021
CAS 1446790-62-0
Color White
Purity Grade Notes Research
Molecular Formula C18H25N7O
Quantity 10 mg
Solubility Information DMSO : 3.03 mg/mL (8.52 mM; Need ultrasonic)
SMILES NC(C1=C(NC2=CC=C(C)C(C)=N2)C=C(N[C@H]3[C@@H](N)CCCC3)N=N1)=O
Molecular Weight (g/mol) 355.44
Formula Weight 355.44
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation