missing translation for 'onlineSavingsMsg'
Learn More

RO8994, MedChemExpress

Product Code. p-200102116
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16497216 100 mg 100mg
16489777 5 mg 5mg
16483267 50 mg 50mg
16427258 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16497216

Brand: Medchem Express HY16999

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RO8994 is a highly potent and selective series of spiroindolinone small-molecule MDM2 inhibitor, with IC50 of 5 nM (HTRF binding assays) and 20 nM (MTT proliferation assays).

RO8994 is a highly potent and selective series of spiroindolinone small-molecule MDM2 inhibitor, with IC50 of 5 nM (HTRF binding assays) and 20 nM (MTT proliferation assays).

Specifications

Chemical Name or Material RO8994
CAS 1309684-94-3
Color White
Purity Grade Notes Research
Molecular Formula C31H31Cl2FN4O4
Quantity 100 mg
Solubility Information DMSO : ≥ 45 mg/mL (73.35 mM)
SMILES O=C([C@H](N[C@H]1CC(C)(C)C)[C@H](C2=CC=CC(Cl)=C2F)[C@@]31C(NC4=C3C=CC(Cl)=C4)=O)NC5=CC=C(C(N)=O)C=C5OC
Molecular Weight (g/mol) 613.51
Formula Weight 613.51
Percent Purity 98.95%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.