missing translation for 'onlineSavingsMsg'
Learn More

RO4987655, MedChemExpress

Product Code. p-200097265
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16488737

Brand: Medchem Express HY14719

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RO4987655 is an orally active and highly selective MEK inhibitor with an IC50 of 5.2 nM for inhibition of MEK1/MEK2.

RO4987655 is an orally active and highly selective MEK inhibitor with an IC50 of 5.2 nM for inhibition of MEK1/MEK2.

Specifications

Chemical Name or Material RO4987655
CAS 874101-00-5
Color White
Purity Grade Notes Research
Molecular Formula C20H19F3IN3O5
Quantity 5 mg
Synonym CH4987655
Solubility Information DMSO : 100 mg/mL (176.90 mM; Need ultrasonic)
SMILES FC(C(F)=C(NC1=CC=C(I)C=C1F)C(C(NOCCO)=O)=C2)=C2CN3C(CCCO3)=O
Molecular Weight (g/mol) 565.28
Formula Weight 565.28
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.