missing translation for 'onlineSavingsMsg'
Learn More

Ro 67-7476, MedChemExpress

Product Code. 16435737
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16435737 5 mg 5mg
16444306 100 mg 100mg
16422998 10 mg 10mg
16419566 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16435737

Brand: Medchem Express HY100403

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ro 67-7476 is a potent positive allosteric modulator of mGluR1 and potentiates glutamate-induced calcium release in HEK293 cells expressing rat mGluR1a with an EC50 of 60.1 nM. Ro 67-7476 is a potent P-ERK1/2 agonist and activates ERK1/2 phosphorylation in the absence of exogenously added glutamate (EC50=163.3 nM).

Ro 67-7476 is a potent positive allosteric modulator of mGluR1 and potentiates glutamate-induced calcium release in HEK293 cells expressing rat mGluR1a with an EC50 of 60.1 nM. Ro 67-7476 is a potent P-ERK1/2 agonist and activates ERK1/2 phosphorylation in the absence of exogenously added glutamate (EC50=163.3 nM).

Specifications

Chemical Name or Material Ro 67-7476
CAS 298690-60-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H18FNO2S
Quantity 5 mg
Solubility Information DMSO : ≥ 40 mg/mL (125.24 mM)
SMILES O=S(N1[C@H](C2=CC=C(F)C=C2)CCC1)(C3=CC=C(C)C=C3)=O
Molecular Weight (g/mol) 319.39
Formula Weight 319.39
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.