missing translation for 'onlineSavingsMsg'
Learn More

RNPC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25820-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894375

  • 4593.76 NOK / 150µL
Estimated Shipment: 19-12-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

U11/U12 snRNP 65K, also known as RBM40 (RNA-binding protein 40), RNPC3 (RNA-binding region-containing protein 3) or RNP, is a 517 amino acid nuclear protein that plays a role in pre-mRNA U12-dependent splicing by the minor spliceosome, ultimately leading to the removal of U12-type introns. Expressed at highest levels in kidney and pancreas, U11/U12 snRNP 65K is also weakly detected in placenta, heart, brain, lung, liver, thymus, colon, ovary, testis, prostate, spleen and small intestine. The gene encoding U11/U12 snRNP 65K maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson′s disease, Gaucher disease, schizophrenia and Usher syndrome.
Specifications

Specifications

RNPC3
Polyclonal
Unconjugated
RNPC3
KIAA183, KIAA1839, RBM40, RNA binding motif protein 40, RNA binding protein 40, RNP, RNPC3, U11/U12 65K, U11/U12 snRNP 65 kDa protein
Rabbit
Antigen Affinity Chromatography
RUO
55599, 67225
-20°C
Liquid
Immunoprecipitation, Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3UZ01, Q96LT9
RNPC3
RNPC3 Fusion Protein Ag22957
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers