missing translation for 'onlineSavingsMsg'
Learn More

RNF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20437-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16849974

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene contains a RING-H2 finger motif. Deletions and mutations in this gene were detected in esophageal squamous cell carcinoma (ESCC), suggesting that this protein may be a potential tumor suppressor. Studies of the mouse counterpart suggested a role of this protein in the transcription regulation that controls germinal differentiation. Multiple alternatively spliced transcript variants encoding the same protein are observed.
Specifications

Specifications

RNF6
Polyclonal
Unconjugated
RNF6
RNF6, SPG2
Rabbit
Antigen Affinity Chromatography
RUO
6049, 74132
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9DBU5, Q9Y252
RNF6
RNF6 Fusion Protein Ag14209
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers