missing translation for 'onlineSavingsMsg'
Learn More

RNF44 Antibody, Novus Biologicals™

Product Code. 18223402 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
18223402 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 18223402

Brand: Novus Biologicals NBP153143

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNF44 Polyclonal specifically detects RNF44 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF44
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q7L0R7
Gene Alias KIAA1100, ring finger protein 44
Gene Symbols RNF44
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RNF44(ring finger protein 44) The peptide sequence was selected from the N terminal of RNF44. Peptide sequence LSYTVTTVTTQGFPLPTGQHIPGCSAQQLPACSVMFSGQHYPLCCLPPPL.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22838
Test Specificity Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Human: 100%; Pig: 100%; Bovine: 92%; Equine: 92%; Mouse: 92%; Rat: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.