Learn More
RNF219 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24988-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
RNF219 (Ring Finger Protein 219, C13orf7) is a Protein Coding gene.Specifications
RNF219 | |
Polyclonal | |
Unconjugated | |
RNF219 | |
C13orf7, ring finger protein 219, RNF219 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79596 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5W0B1 | |
RNF219 | |
RNF219 Fusion Protein Ag21767 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |