missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF213 Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18409JF525
This item is not returnable.
View return policy
Description
RNF213 Polyclonal antibody specifically detects RNF213 in Human samples. It is validated for Western Blot
Specifications
| RNF213 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 525 | |
| ALK lymphoma oligomerization partner on chromosome 17, C17orf27, chromosome 17 open reading frame 27, DKFZp762N1115, FLJ13051, KIAA1554MGC46622, KIAA1618, MGC9929, NET57, protein ALO17, ring finger protein 213 | |
| Recognizes region between residues Between between 4300 and 4350.. | |
| 0.1 mL | |
| Zinc Finger | |
| 57674 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction