Learn More
RNF145 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24524-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The RNF145 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human RNF145. This antibody recognizes human antigen. The RNF145 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
RNF145 | |
Polyclonal | |
Unconjugated | |
20 μL | |
153830 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human |