Learn More
RNF145 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24524-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16803005
  • 1611.84 NOK / 20µL
Estimated Shipment: 21-05-2024
Description
The RNF145 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human RNF145. This antibody recognizes human antigen. The RNF145 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
RNF145
Polyclonal
Unconjugated
20 μL
153830
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
367 μg/mL