Learn More
Abnova™ RND3 (Human) Recombinant Protein
Human RND3 full-length ORF ( AAH12513, 1 a.a. - 244 a.a.) recombinant protein with GST-tag at N-terminal.
Brand:  Abnova™ H00000390-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16131841
  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
Description
  • Sequence: MKERRASQKLSSKSIMDPNQNVKCKIVVVGDSQCGKTALLHVFAKDCFPENYVPTVFENYTASFEIDTQRIELSLWDTSGSPYYDNVRPLSYPDSDAVLICFDISRPETLDSVLKKWKGEIQEFCPNTKMLLVGCKSDLRTDVSTLVELSNHRQTPVSYDQGANMAKQIGAATYIECSALQSENSVRDIFHVATLACVNKTNKNVKRNKSQRATKRISHMPSRPELSAVATDLRKDKAKSCTVM
Specifications
Specifications
AAH12513
Rho family GTPase 3
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RND3
GST
390
Wheat germ expression system
25 μg
ARHE, Rho8, RhoE, memB
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer