Learn More
MP Biomedicals™ RNase-Erase Decontaminants
Product Code. p-83822 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (Metric):
250 mL
2 x 125 mL
Packaging Type:
Dropper Bottles
Spray Bottle
Unit Size:
250mL
Product Code. Volume (Metric) Packaging Type unitSize
11421722 250 mL Spray Bottle 250mL
11431722 2 x 125 mL Dropper Bottles 250mL
2 options
Product Code. 11421722
Brand: MP Biomedicals™ 04821682
Completely removes RNase contamination from glass and plastic surfaces
Eliminates the need to bake glassware or use DEPC.
  • Leaves no residues that might inhibit enzymatic reactions
  • Supplied as ready-to-use formula
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 250 mL
Packaging Type Spray Bottle