Learn More
Invitrogen™ RNAqueous™ Total RNA Isolation Kit
Product Code. 10176864
Change view
Click to view available options
Quantity:
50 Catridges
50 Preps
Product Type:
RNA Isolation Kit
Spin Columns
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Unit Size
10176864 50 Catridges Spin Columns Each
10075994 50 Preps RNA Isolation Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10176864 Supplier Invitrogen™ Supplier No. AM10051G
  • ONLINE EXCLUSIVE PRICE
    2380.00 NOK
    2308.65 NOK / Pack of 50
    Save 71.35 NOK
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 10^2–10^7 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.
The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 102–107 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.
The RNAqueous kit provides:
  • Convenient syringe-based filtration
  • Compatibility with vacuum manifolds when processing multiple samples
How it works
The RNAqueous kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 50 cartridges; store at room temperature.
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product Type Spin Columns
Quantity 50 Catridges
Shipping Condition Room Temperature