Learn More
Invitrogen™ Rat Set
Description
Stealth RNAi Predesigned siRNAs enable robust and long-lasting gene silencing with reduced off-target effects. Stealth RNAi siRNAs are modified synthetic 25-mer RNA duplexes. Our proprietary technology helps ensure high specificity and stability, making Stealth RNAi siRNAs an excellent choice for researchers seeking reliable and efficient gene knockdown in human, mouse, or rat.
Key features include:
Higher specificity and potency—algorithm designs minimize off-target effects and maximize target gene knockdown
Enhanced stability—engineered for enhanced stability in serum and cellular environments, ensuring prolonged gene silencing
Minimal cellular toxicity—designed to evade immune detection, for reduced risk of immune response
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Application) | RNAi |
| Product Type | siRNA |
| Quantity | 1 Set |
| Target | Human Set |