missing translation for 'onlineSavingsMsg'
Learn More

RN-1734, MedChemExpress

Product Code. p-200101180
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16483917 50 mg 50mg
16450458 5 mg 5mg
16427776 100 mg 100mg
16417998 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16483917

Brand: Medchem Express HY19975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RN-1734 is selective antagonist of the TRPV4 channel, completely antagonizes 4αPDD-mediated activation of TRPV4 with comparable, low micromolar IC50s for all three species (hTRPV4: 2.3 μM, mTRPV4: 5.9 μM, rTRPV4: 3.2 μM). RN-1734 clearly decreases the production of tumor necrosis factor α (TNF-α) and interleukin 1β (IL-1β) without altering the number of olig2-positive cells.

RN-1734 is selective antagonist of the TRPV4 channel, completely antagonizes 4αPDD-mediated activation of TRPV4 with comparable, low micromolar IC50s for all three species (hTRPV4: 2.3 μM, mTRPV4: 5.9 μM, rTRPV4: 3.2 μM). RN-1734 clearly decreases the production of tumor necrosis factor α (TNF-α) and interleukin 1β (IL-1β) without altering the number of olig2-positive cells.

Specifications

Chemical Name or Material RN-1734
CAS 946387-07-1
Color White
Purity Grade Notes Research
Molecular Formula C14H22Cl2N2O2S
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (70.76 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C1=CC=C(Cl)C=C1Cl)(N(C(C)C)CCNC(C)C)=O
Molecular Weight (g/mol) 353.31
Formula Weight 353.31
Percent Purity 98.99%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.