Learn More
RN-1734, MedChemExpress
Product Code. 16483917
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16483917 50 mg 50mg
16450458 5 mg 5mg
16427776 100 mg 100mg
16417998 10 mg 10mg
4 options
Product Code. 16483917
Brand: Medchem Express HY19975
MedChemExpress RN-1734 is selective antagonist of the TRPV4 channel, completely antagonizes 4αPDD-mediated activation of TRPV4 with comparable, low micromolar IC50s for all three species (hTRPV4: 2.3 μM, mTRPV4: 5.9 μM, rTRPV4: 3.2 μM). RN-1734 clearly decreases the production of tumor necrosis factor α (TNF-α) and interleukin 1β (IL-1β) without altering the number of olig2-positive cells.
RN-1734 is selective antagonist of the TRPV4 channel, completely antagonizes 4αPDD-mediated activation of TRPV4 with comparable, low micromolar IC50s for all three species (hTRPV4: 2.3 μM, mTRPV4: 5.9 μM, rTRPV4: 3.2 μM). RN-1734 clearly decreases the production of tumor necrosis factor α (TNF-α) and interleukin 1β (IL-1β) without altering the number of olig2-positive cells.
Specifications
Chemical Name or Material RN-1734
CAS 946387-07-1
Color White
Purity Grade Notes Research
Molecular Formula C14H22Cl2N2O2S
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (70.76 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C1=CC=C(Cl)C=C1Cl)(N(C(C)C)CCNC(C)C)=O
Molecular Weight (g/mol) 353.31
Formula Weight 353.31
Percent Purity 98.99%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation