missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RN-1734, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
RN-1734 is selective antagonist of the TRPV4 channel, completely antagonizes 4αPDD-mediated activation of TRPV4 with comparable, low micromolar IC50s for all three species (hTRPV4: 2.3 μM, mTRPV4: 5.9 μM, rTRPV4: 3.2 μM). RN-1734 clearly decreases the production of tumor necrosis factor α (TNF-α) and interleukin 1β (IL-1β) without altering the number of olig2-positive cells.
Specifications
Specifications
| Chemical Name or Material | RN-1734 |
| CAS | 946387-07-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H22Cl2N2O2S |
| Quantity | 10 mg |
| Solubility Information | DMSO : 25 mg/mL (70.76 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=S(C1=CC=C(Cl)C=C1Cl)(N(C(C)C)CCNC(C)C)=O |
| Molecular Weight (g/mol) | 353.31 |
| Formula Weight | 353.31 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction