missing translation for 'onlineSavingsMsg'
Learn More

RMC-4550, MedChemExpress

Product Code. p-200098511
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16464858

Brand: Medchem Express HY116009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RMC-4550 is a potent, selective and allosteric inhibitor of SHP2, with an IC50 of 0.583 nM.

RMC-4550 is a potent, selective and allosteric inhibitor of SHP2, with an IC50 of 0.583 nM.

Specifications

Chemical Name or Material RMC-4550
CAS 2172651-73-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H26Cl2N4O2
Quantity 10 mg
Solubility Information DMSO : ≥ 260 mg/mL (594.48 mM)
SMILES ClC1=C(C(C2=NC(CO)=C(N=C2C)N3CCC4(CC3)CO[C@H]([C@H]4N)C)=CC=C1)Cl
Molecular Weight (g/mol) 437.36
Formula Weight 437.36
Percent Purity 98.61%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.