Learn More
Rivastigmine impurity 1, TRC
Specifications
Specifications
| Chemical Name or Material | Rivastigmine impurity 1 |
| CAS | 1369779-38-3 |
| Molecular Formula | C10 H15 N O2 |
| Quantity | 250 mg |
| Synonym | 3-[(1S)-1-(Dimethyloxidoamino)ethyl]phenol, (S)-1-(3-hydroxyphenyl)-N,N-dimethylethan-1-amine oxide(S)-1-(3-hydroxyphenyl)-N,N-dimethylethan-1-amine oxide |
| InChI Formula | InChI=1S/C10H15NO2/c1-8(11(2,3)13)9-5-4-6-10(12)7-9/h4-8,12H,1-3H3/t8-/m0/s1 |
| SMILES | CN([C@H](C1=CC(O)=CC=C1)C)(C)=O |
| Molecular Weight (g/mol) | 181.23 |