Learn More
Rivaroxaban, 98%
Product Code. 15517739 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15517739 1 g 1g
15527739 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15517739 Supplier Thermo Scientific Acros Supplier No. 461600010
Request Bulk or Custom Format
CAS: 366789-02-8 | C19H18ClN3O5S | 435.88 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 366789-02-8
Molecular Formula C19H18ClN3O5S
Molecular Weight (g/mol) 435.88
MDL Number MFCD11974010
InChI Key KGFYHTZWPPHNLQ-UHFFFAOYNA-N
Synonym rivaroxaban, xarelto, s-5-chloro-n-2-oxo-3-4-3-oxomorpholino phenyl oxazolidin-5-yl methyl thiophene-2-carboxamide, unii-9ndf7jz4m3, xarelto tn, 9ndf7jz4m3, 5-chloro-n-5s-2-oxo-3-4-3-oxomorpholin-4-yl phenyl-1,3-oxazolidin-5-yl methyl thiophene-2-carboxamide, 2-thiophenecarboxamide, 5-chloro-n-5s-2-oxo-3-4-3-oxo-4-morpholinyl phenyl-5-oxazolidinyl methyl, 5-chloro-n-5s-2-oxo-3-4-3-oxo-4-morpholinyl phenyl-5-oxazolidinyl methyl-2-thiophenecarboxamide
PubChem CID 9875401
ChEBI CHEBI:68579
IUPAC Name 5-chloro-N-[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl]thiophene-2-carboxamide
SMILES ClC1=CC=C(S1)C(=O)NCC1CN(C(=O)O1)C1=CC=C(C=C1)N1CCOCC1=O
Specifications
Melting Point 228.0°C to 229.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 435.88
Percent Purity 98%
Chemical Name or Material Rivaroxaban
compliance-icons compliance-icons
Product Identifier
  • Rivaroxaban
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste d.