missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rituximab Rabbit anti-Multi-Species, Clone: 2260A, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9630-100
This item is not returnable.
View return policy
Description
Rituximab Monoclonal specifically detects Rituximab in Multi-Species samples. It is validated for Western Blot, Flow Cytometry, ELISA, Binding Inhibition.Specifications
| Rituximab | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Binding Inhibition | |
| 2260A | |
| Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA, Binding Inhibition 2.5 ug/mL | |
| B lymphocyte antigen CD20, B lymphocyte surface antigen B1, Bp35, CD20, LEU 16, LEU16, Membrane spanning 4 domains subfamily A member 1, MS4A1 | |
| Full-length Rituximab | |
| 100 μg | |
| Detects Rituximab in Western blots in direct ELISAs. 100% cross-reactivity with Tositumomab and Ibritumomab (95% or higher sequence identity with Rituximab) and no cross-reactivity with Cetuximab, Trastuzumab, Brentuximab, Gemtuzumab, Adalimumab, Solanezumab, Teprotumumab, and Aducanumab is observed. | |
| Multi-Species | |
| Supernatant |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction