Learn More
Ritter Plastic™ Ritips™ Dispenser Tips
Used for the repetitive distribution of volumes between 0.05 and 50 mL.
Brand:  Ritter Plastic™ 40010-0002
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16610302
  • 25508.90 NOK / Pack of 3000
Estimated Shipment: 15-07-2024
Description
  • Uses direct displacement method
  • Works well for infectious and volatile liquids
  • Fluid never in touch with the instrument to avoid contamination
  • Automated individual tests of each single tip guarantee the tightness with water, which is documented in lot-specific records
  • Black graduation marks
  • Manufactured from polypropylene
  • Suitable for a wide range of pipettors
Specifications
Specifications
1.25 mL
100/Pk.
Yes
Polypropylene
Pipetting
Wide range of pipettors
RiTips