Learn More
RIPK3/RIP3 [p Ser232] Antibody (JE44-62), Novus Biologicals™
Product Code. 30115267 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30115267 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30115267 Supplier Novus Biologicals Supplier No. NBP332795
Rabbit Monoclonal Antibody
RIPK3/RIP3 Monoclonal antibody specifically detects RIPK3/RIP3 in Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Antigen RIPK3/RIP3
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone JE44-62
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
Formulation 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol
Gene Alias EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser232 of Human RIPK3/RIP3. (Uniprot: Q9QZL0)
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11035
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG