Learn More
Eurofins DiscoverX RIPK2 Protein, Active
Description
RIPK2 (receptor-interacting serine-threonine kinase 2, RIP2, RICK, CARD3, CARDIAK) is a death domain-containing serine-threonine kinase associated with TNF receptor and implicated in the activation of NF-kappaB and cell death in mammalian cells. RIPK2 represents a kinase that regulates TNF and Fas induced-apoptosis and that is involved in the generation of proinflammatory cytokine IL-1β.
GenBank NM_003821
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 37.8 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥59% |
| Protein | RIPK2 Kinase, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.