missing translation for 'onlineSavingsMsg'
Learn More

RIOK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen RIOK3
Concentration 0.15 mg/mL
Applications Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16841884
View Documents
Proteintech
13593-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16831884
View Documents
Proteintech
13593-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.
Specifications

Specifications

RIOK3
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
O14730, Q9DBU3
361293, 66878, 8780
RIOK3 Fusion Protein Ag4497
Primary
-20°C
RIOK3
0.15 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
RIO kinase 3, RIO kinase 3 (yeast), RIOK3, SUDD, sudD homolog
Riok3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.