Learn More
Riociguat, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Riociguat
Specifications
Specifications
| Chemical Name or Material | Riociguat |
| CAS | 625115-55-1 |
| Analyte or Component Names | Riociguat |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H19 F N8 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Carbamic acid, N-[4,6-diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-pyrimidinyl]-N-methyl-, methyl ester, Carbamic acid, [4,6-diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-pyrimidinyl]methyl-, methyl ester (9CI), Adempas, BAY 63-2521, BAY 632521, Methyl [4,6-diamino-2-[1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-5-yl]methylcarbamate |
| InChI Formula | InChI=1S/C20H19FN8O2/c1-28(20(30)31-2)15-16(22)25-18(26-17(15)23)14-12-7-5-9-24-19(12)29(27-14)10-11-6-3-4-8-13(11)21/h3-9H,10H2,1-2H3,(H4,22,23,25,26) |
| SMILES | COC(=O)N(C)c1c(N)nc(nc1N)c2nn(Cc3ccccc3F)c4ncccc24 |
| IUPAC Name | methyl N-[4,6-diamino-2-[1-[(2-fluorophenyl)methyl]pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-5-yl]-N-methylcarbamate |
| Show More |