Learn More
Ansell Ringers R074 PVC-Coated Chemical Resistant/Waterproof Impact Gloves
Product Code. 16689063 Shop All Ansell Products
Click to view available options
Size:
Small
Large
2X-Large
Unit Size:
12 pairs
Product Code. 16689063
Brand: Ansell 07410
Only null left
Add to basket
Add to basket
Developed with Ringers' F3 Technology™ and patented TPR design for increased dexterity and comfort. Textured PVC coating on the palm enhances grip capabilities, while also providing chemical resistance.
  • TPR impact protection on top of the hand and over the full length of the fingers
  • PVC coating is waterproof and provides resistance to chemicals
  • Inner liner provides cut resistance throughout the entire glove
  • Durable textured palm coating enhances grip capabilities
  • Short gauntlet-style cuff keeps debris away from the hands
  • Level 1 Contact Heat performance rating at an outside exposure of 100°C, in accordance with test method EN 407
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) EN 374_2003 Microorganism, EN 420:2003 + A1:2009
Applications Industrial
Coating Material PVC
Color Hi-vis Red
Cut Resistance Rating ANSI A3
Exterior Finish Fully Textured
Latex Free Yes
Material DuPont™ Aramid Fibers
Product Type Chemical-resistant Gloves
Size Large
EN388 Standard 4X41CP
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMO
EN ISO 374 - 5 Yes
Glove Cuff Style Gauntlet
Series R-074
Length (Metric) 355 mm