Description
- TPR impact protection on top of the hand and over the full length of the fingers
- PVC coating is waterproof and provides resistance to chemicals
- Inner liner provides cut resistance throughout the entire glove
- Durable textured palm coating enhances grip capabilities
- Short gauntlet-style cuff keeps debris away from the hands
- Level 1 Contact Heat performance rating at an outside exposure of 100°C, in accordance with test method EN 407
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 5 |
| Certifications/Compliance (Europe) | EN 374_2003 Microorganism, EN 420:2003 + A1:2009 |
| Applications | Industrial |
| Coating Material | PVC |
| Color | Hi-vis Red |
| Cut Resistance Rating | ANSI A3 |
| Exterior Finish | Fully Textured |
| Latex Free | Yes |
| Material | DuPont™ Aramid Fibers |
| Product Type | Chemical-resistant Gloves |
| Show More |