Learn More
Ansell Ringers 065 R-Flex Impact Glove
Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
12
13
Unit Size:
12 pairs
Description
- The Ringers 065 R-Flex Impact glove with a Nitrile Half Dip palm combines protection against impact hazards with cut resistance for a wide range of applications.
- A breathable knit shell developed with Ringers' F3 Technology™ and patented TPR design provide superior dexterity and comfort.
- The sandy finish palm ensures secure grip in wet and dry environments, and touchscreen compatible index, middle and thumb tips allow technology interaction.
Specifications
Specifications
| Coating Material | Nitrile |
| Color | Hi-vis Yellow, Gray |
| Cut Resistance Rating | ANSI A4 |
| Hand | Specific |
| Latex Free | Yes |
| Material | HPPE |
| Product Type | Mechanical Protection Gloves |
| Size | 9 |
| Glove Cuff Style | Knit |