Learn More
Rimegepant, MedChemExpress
Product Code. 16469147
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16469147 5 mg 5mg
16462667 50 mg 50mg
16456578 10 mg 10mg
16426786 100 mg 100mg
4 options
Product Code. 16469147
Brand: Medchem Express HY15498
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MedChemExpress Rimegepant (BMS-927711) is a highly potent, oral calcitonin gene-related peptide (CGRP) receptor antagonist with a Ki of 0.027 nM and an IC50 of 0.14 nM for hCGRP receptor.
Rimegepant (BMS-927711) is a highly potent, oral calcitonin gene-related peptide (CGRP) receptor antagonist with a Ki of 0.027 nM and an IC50 of 0.14 nM for hCGRP receptor.
Specifications
Chemical Name or Material Rimegepant
CAS 1289023-67-1
Color White
Purity Grade Notes Research
Molecular Formula C28H28F2N6O3
Quantity 5 mg
Synonym BMS-927711
Solubility Information DMSO : 10 mg/mL (18.71 mM; Need ultrasonic and warming) ∣Ethanol : 4.4 mg/mL (8.23 mM; Need ultrasonic)
SMILES N[C@H]1[C@H](C2=C(F)C(F)=CC=C2)CC[C@@H](OC(N3CCC(N4C(C=CC=N5)=C5NC4=O)CC3)=O)C6=NC=CC=C61
Molecular Weight (g/mol) 534.56
Formula Weight 534.56
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation