Learn More
Brady™ Rigid Polypropylene Prohibition Signs
Specifications
Specifications
| Certifications/Compliance | NF X 08-003-1 and 08-003-3 NX and the European Directive 92/58 / EEC |
| Material | Polypropylene |
| Quantity | 1 Ea. |
| Description | Rigid Polypropylene |
| Diameter (Metric) | 315 mm |
| For Use With (Application) | Used for a wide variety of applications both indoors and outdoors |
| Max. Temperature (Metric) | 120°C |
| Width (Metric) | 105 mm |
| Shape | Round |
| Displayed Text | Defense to use cell phones |
| Show More |