Learn More
Brady™ Rigid Polypropylene Evacuation Panel Safety Signs
Specifications
Specifications
| Material | Polypropylene |
| Quantity | 1 Ea. |
| Description | Rigid Polypropylene |
| Height (Metric) | 210 mm |
| Max. Temperature (Metric) | 120°C |
| Thickness | 0015 mm |
| Width (Metric) | 15 mm |
| Displayed Text | Exit and Emergency Exit Down Arrow |
| Product Type | Safety Sign |
| Color | White on Green |