Learn More
Rifaximin, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 80621-81-4 |
| Molecular Formula | C43H51N3O11 |
| MDL Number | MFCD00864973 |
| Quantity | 1 g |
| SMILES | C[C@H]1/C=C/C=C(/C(=O)NC2=C(C3=C(C4=C(C(=C3O)C)O[C@@](C4=O)(O/C=C/[C@@H]([C@H]([C@H]([C@@H]([C@@H]([C@@H]([C@H]1O)C)O)C)OC(=O)C)C)OC)C)C5=C2N6C=CC(=CC6=N5)C)O)\C |
| Molecular Weight (g/mol) | 785.88 |
| Percent Purity | 98% |
| Chemical Name or Material | Rifaximin |