Learn More
Rifamycin SV Sodium, TRC
Specifications
Specifications
| Chemical Name or Material | Rifamycin sodium |
| CAS | 14897-39-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C37 H46 N O12 . Na |
| Quantity | 100 mg |
| Synonym | Rifamycin sodium, Sodium (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-21-(acetyloxy)-6,9,17,19-tetrahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-1,11-dioxo-1,2-dihydro-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-5-olate, 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione','5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-','21-acetate','sodium salt (8CI), Rifamycin','monosodium salt (9CI), 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan','rifamycin deriv., CB 01-11, Monosodium rifamycin SV, NSC 146718, Rifamycin SV sodium, Rifamycin SV sodium salt, Rifamycin sodium, Rifamycin sodium salt, Tuborin, Rifaximin Imp C (EP) |
| InChI Formula | InChI=1S/C37H47NO12.Na/c1-16-11-10-12-17(2)36(46)38-23-15-24(40)26-27(32(23)44)31(43)21(6)34-28(26)35(45)37(8,50-34)48-14-13-25(47-9)18(3)33(49-22(7)39)20(5)30(42)19(4)29(16)41;/h10-16,18-20,25,29-30,33,40-44H,1-9H3,(H,38,46);/q;+1/p-1/b11-10+,14-13+,17-12-;/t16-,18+,19+,20+,25-,29-,30+,33+,37-;/m0./s1 |
| SMILES | [Na+].CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c([O-])c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C)cc(O)c4c3C2=O |
| IUPAC Name | sodium;(7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-13-acetyloxy-2,15,17,29-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.14,7.05,28]triaconta-1(29),2,4,9,19,21,25,27-octaen-27-olate |
| Molecular Weight (g/mol) | 719.75 |
| Show More |