Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
Product Code. 10425339 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Solution A
Solution B
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10425339 Solution A Each
10570519 Solution B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10425339 Supplier Epredia™ Supplier No. 3313
  • ONLINE EXCLUSIVE PRICE
    442.00 NOK
    343.95 NOK / Pack of 1
    Save 98.05 NOK
    22% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Includes
A package insert is included with easy-to-follow instructions.
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
  • Stain can be used to detect H. Pylori.
  • Set contains easy-to-follow instructions
  • Product is available in one-gallon containers to facilitate use on automated strainers
  • Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)
Order Info
Components are also available in one-gallon containers to facilitate use on automated stainers.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 gal.
For Use With (Application) Stain set provides results equal to wright-giemsa
Includes A package insert is included with easy-to-follow instructions
Product Type Solution A