Learn More
Epredia™ Richard-Allan Scientific™ Signature Series Eosin-Y 7111
Product Code. p-4530131 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
Unit Size:
0.47 L
Product Code. Quantity unitSize
12677756 1 pt. 0.47 L
1 options
Product Code. 12677756
Brand: Epredia™ 7111
Create an ideal visual contrast to Hematoxylin with the fast-acting Epredia™ Richard-Allan Scientific™ Signature Series Eosin-Y counterstain.
  • Bright, vivid cytoplasmic staining
  • Creates three distinct shades of pink
  • Unmatched color contrast between the nuclei and cytoplasm
  • Various shades of red and pink differentiate muscle, red blood cells and connective tissue
  • Strict quality assurance guarantees product consistency and longevity
  • Quantitative testing eliminates need for adjustments in dilution or time
Available in 5 liter packaging
  • Reduces plastic by 90%
  • Reduces stain spills
  • Increases stain stability
  • Saves 35% storage space versus traditional packaging
TRUSTED_SUSTAINABILITY
Specifications
Type Eosin Stain
Quantity 1 pt.
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.