Learn More
Epredia™ Richard-Allan Scientific™ Differentiating Solution Product Code.: 12523627
Epredia™ Richard-Allan Scientific™ Differentiating Solution
Product Code. p-4532408 Shop All Epredia Products
1 gal., 3.8 L
Click to view available options
Quantity:
1 gal.
Unit Size:
3.8 L
Product Code. 12523627
Brand: Epredia™ 74211
Obtain clear differentiation and contrast between the hematoxylin and eosin with Epredia™ Richard-Allan Scientific™ Differentiating Solution.
  • Reproducible batch results allow for consistent staining in regressive procedures
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hydrochloric Acid Solution
Quantity 1 gal.
Product Content Correction
Product Title
Epredia™ Richard-Allan Scientific™ Differentiating Solution >