Learn More
Epredia™ Cytoseal™ Mountant
Product Code. 12502736 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 oz.
16 oz.
Product Line:
Cytoseal 60
Cytoseal XYL
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Line unitSize
12502736 4 oz. Cytoseal XYL Pack of 12
11932365 16 oz. Cytoseal 60 Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12502736 Supplier Epredia™ Supplier No. 83124
Prevent cracking, fading and discoloration of stained specimens with Cytoseal™ mounting media.
Cytoseal XYL mountant is a xylene-based rapid drying medium that allows for microscopic examination of slides soon after application.
Ordering Information:
Packaged in an easy-to-use dispenser with a twist-open/twist-closed cap.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Type 4 oz Dispenser Bottle
Product Line Cytoseal XYL
Product Type Mountant
Quantity 4 oz.
Formulation Fast-drying formula reduces waiting time and increases throughput
For Use With (Application) Provides exceptional clarity and longevity for your microscopy slides
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cytoseal XYL
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P201-Obtain special instructions before use.